Gene Transcripts

Gene Symbol
Krtap5-5
Entrez Gene ID
114666
Gene Name
keratin associated protein 5-5
Gene Aliases
A030001H12Rik, AI586182, BB149782
Chromosome Location
Chr. 7: 142228795 - 142229971 on Build GRCm38
UniGene ID
Mm.444495
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037822.1 NP_001032911.1 1 474
GenBank mRNA AK020690.1 - 1 481
BC111113.1 - 1 474 40051363

View Full Product Details