Gene Transcripts
- Gene Symbol
- Olfr74
- Entrez Gene ID
- 117005
- Gene Name
- olfactory receptor 74
- Gene Aliases
- MOR174-4, mOR-EV
- Chromosome Location
- Chr. 2: 87973707 - 87974663 on Build GRCm38
- UniGene ID
- Mm.196679
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_054091.2 |
NP_473432.2
|
1 |
896 |
|
View Full Product Details