Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001177845.1 | NP_001171316.1 | 6 | 1177 | |
NM_001177846.1 | NP_001171317.1 | 6 | 1159 | ||
NM_001177847.1 | NP_001171318.1 | 6 | 1105 | ||
NM_054096.2 | NP_473437.2 | 6 | 1169 | ||
GenBank mRNA | AF378130.1 | - | 6 | 1078 | |
AF410784.1 | - | 5 | 1001 | ||
AK036431.1 | - | 6 | 1098 | ||
AK080135.1 | - | 6 | 1178 | ||
AK151117.1 | - | 6 | 1107 | ||
AK168494.1 | - | 6 | 1097 | ||
BC005584.1 | - | 3 | 835 | 3157913 |