Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289643.1 | NP_001276572.1 | 1 | 158 | |
NM_001289644.1 | NP_001276573.1 | 1 | 158 | ||
GenBank mRNA | AF274959.1 | - | 1 | 132 | |
AF274960.1 | - | 1 | 132 | ||
AF274961.1 | - | 1 | 132 | ||
AF439546.1 | - | 1 | 124 | ||
AK136373.1 | - | 1 | 136 | ||
AK136396.1 | - | 1 | 135 | ||
BC061046.1 | - | 132 | 30301032 | ||
BY120182.1 | - | 1 | 159 |