Gene Transcripts

Gene Symbol
Ubox5
Entrez Gene ID
140629
Gene Name
U box domain containing 5
Gene Aliases
1500010O06Rik, C330018L13Rik, Rnf37, UIP5, Ubce7ip5, Ube7ip5
Chromosome Location
Chr. 2: 130589996 - 130630038 on Build GRCm38
UniGene ID
Mm.486003
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001255993.1 NP_001242922.1 5 1191
NM_001255994.1 NP_001242923.1 4 1044
NM_080562.5 NP_542129.2 4 1049
GenBank mRNA AF360997.1 - 4 979
AK034114.1 - 4 998
AK049276.1 - 3 815
AK158052.1 - 5 1149
AK170024.1 - 5 2892
AK170100.1 - 4 1020
AK171070.1 - 5 1158
BC025068.1 - 3 911 4242459

View Full Product Details