Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_130450.2 | NP_569717.1 | 2 | 195 | |
GenBank mRNA | AB072039.1 | - | 2 | 142 | |
AF480860.1 | - | 2 | 114 | ||
AK029029.1 | - | 2 | 197 | ||
AK087440.1 | - | 2 | 215 | ||
AK165847.1 | - | 1 | 219 | ||
AK167373.1 | - | 2 | 161 | ||
AY053453.1 | - | 2 | 176 | ||
BC040097.1 | - | 2 | 137 | ||
BC051041.1 | - | 2 | 192 | 4012676 | |
BC098492.1 | - | 1 | 192 | 30091954 | |
BC100576.1 | - | 2 | 137 | 30097278 |