Gene Transcripts

Gene Symbol
Usp48
Entrez Gene ID
170707
Gene Name
ubiquitin specific peptidase 48
Gene Aliases
2810449C13Rik, AI115503, BC021769, D330022K21Rik, Usp31
Chromosome Location
Chr. 4: 137593757 - 137658537 on Build GRCm38
UniGene ID
Mm.218478
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130879.2 NP_570949.2 12 1792
GenBank mRNA AK034556.1 - 1 2740
AK052297.1 - 12 1734
AK133251.1 - 13 1809
BC021769.1 - 12 1753

View Full Product Details