Gene Transcripts

Gene Symbol
Usp48
Entrez Gene ID
170707
Gene Name
ubiquitin specific peptidase 48
Gene Aliases
2810449C13Rik, AI115503, BC021769, D330022K21Rik, Usp31
Chromosome Location
Chr. 4: 137593757 - 137658537 on Build GRCm38
UniGene ID
Mm.218478
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130879.2 NP_570949.2 18, 19 2457
GenBank mRNA AK052297.1 - 18, 19 2399
AK133251.1 - 19, 20 2474
BC058410.1 - 5, 6 500

View Full Product Details