Gene Transcripts
- Gene Symbol
- Otud7a
- Entrez Gene ID
- 170711
- Gene Name
- OTU domain containing 7A
- Gene Aliases
- AJ430384, Otud7
- Chromosome Location
- Chr. 7: 63444745 - 63759905 on Build GRCm38
- UniGene ID
- Mm.156872
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_130880.1 |
NP_570950.1
|
14 |
1870 |
|
GenBank mRNA |
AJ430384.1 |
- |
14 |
1870 |
|
View Full Product Details