Gene Transcripts
- Gene Symbol
- Vmn1r24
- Entrez Gene ID
- 171191
- Gene Name
- vomeronasal 1 receptor 24
- Gene Aliases
- V1rc18
- Chromosome Location
- Chr. 6: 57955641 - 57956531 on Build GRCm38
- UniGene ID
- Mm.377171
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_134173.2 |
NP_598934.1
|
1 |
851 |
|
View Full Product Details