Gene Transcripts

Gene Symbol
Vmn1r70
Entrez Gene ID
171262
Gene Name
vomeronasal 1 receptor 70
Gene Aliases
V1rd2, V1rl1
Chromosome Location
Chr. 7: 10633587 - 10634483 on Build GRCm38
UniGene ID
Mm.222634
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134228.1 NP_598989.1 1 715
GenBank mRNA AF394955.1 - 1 715

View Full Product Details