Gene Transcripts

Gene Symbol
Rnf43
Entrez Gene ID
207742
Gene Name
ring finger protein 43
Gene Aliases
4732452J19Rik
Chromosome Location
Chr. 11: 87662464 - 87735539 on Build GRCm38
UniGene ID
Mm.440230
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172448.3 NP_766036.2 8 1560
GenBank mRNA AK028750.1 - 7 919
BC075707.1 - 6 696
BC131957.1 - 6 678 40130234

View Full Product Details