Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001171801.1 | NP_001165272.1 | 5 | 850 | |
NM_173746.3 | NP_776107.2 | 5 | 847 | ||
GenBank mRNA | AK048604.1 | - | 5 | 826 | |
BC147183.1 | - | 1 | 415 | 9056211 | |
BC147184.1 | - | 1 | 415 | 9056218 |