Gene Transcripts

Gene Symbol
Dock3
Entrez Gene ID
208869
Gene Name
dedicator of cyto-kinesis 3
Gene Aliases
MOCA, PBP, mKIAA0299
Chromosome Location
Chr. 9: 106892825 - 107231909 on Build GRCm38
UniGene ID
Mm.150259
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153413.2 NP_700462.2 17 2019
GenBank mRNA AK147385.1 - 3 332
AY145302.1 - 17 1969
CK781165.1 - 6 706

View Full Product Details