Gene Transcripts

Gene Symbol
Zfyve26
Entrez Gene ID
211978
Gene Name
zinc finger, FYVE domain containing 26
Gene Aliases
4930465A13, 9330197E15Rik, A630028O16Rik, Gm893, mKIAA0321
Chromosome Location
Chr. 12: 79232346 - 79296310 on Build GRCm38
UniGene ID
Mm.297919
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008550.1 NP_001008550.1 15 2751
GenBank mRNA AK041668.1 - 15 2748
AK132344.1 - 15 2741
AK220333.1 - 15 2722
BC141359.1 - 15 2734 9055971

View Full Product Details