Gene Transcripts

Gene Symbol
Inpp5a
Entrez Gene ID
212111
Gene Name
inositol polyphosphate-5-phosphatase A
Gene Aliases
Chromosome Location
Chr. 7: 139389109 - 139579652 on Build GRCm38
UniGene ID
Mm.28308
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127363.1 NP_001120835.1 4 562
NM_183144.3 NP_898967.2 4 562
GenBank mRNA AK083449.1 - 5 561
AK157558.1 - 4 425
AK171360.1 - 4 562
AK171880.1 - 4 538
BC056341.1 - 4 562 5703345

View Full Product Details