Gene Transcripts

Gene Symbol
Frat2
Entrez Gene ID
212398
Gene Name
frequently rearranged in advanced T cell lymphomas 2
Gene Aliases
Chromosome Location
Chr. 19: 41845972 - 41848132 on Build GRCm38
UniGene ID
Mm.222723
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177603.3 NP_808271.1 1 1259
GenBank mRNA AK081171.1 - 1 1262
BC034214.1 - 1 1222 4989784
BC085510.1 - 1 1235 5718518

View Full Product Details