Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145423.2 | NP_663398.2 | 13 | 1859 | |
GenBank mRNA | AK039927.1 | - | 13 | 1688 | |
AK132487.1 | - | 1 | 19 | ||
AK136457.1 | - | 13 | 1869 | ||
AY484428.1 | - | 13 | 1859 | ||
BC017691.1 | - | 1814 | 4239161 |