Gene Transcripts

Gene Symbol
Clptm1l
Entrez Gene ID
218335
Gene Name
CLPTM1-like
Gene Aliases
C130052I12Rik, Crr9
Chromosome Location
Chr. 13: 73604002 - 73620639 on Build GRCm38
UniGene ID
Mm.30099
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146047.2 NP_666159.2 7 1077
GenBank mRNA AK032295.1 - 7 1040
AK048357.1 - 7 974
AK156203.1 - 7 968
AK165939.1 - 7 938
AK177383.1 - 2 66
BC026562.1 - 7 778 5028264

View Full Product Details