Gene Transcripts

Gene Symbol
Cd226
Entrez Gene ID
225825
Gene Name
CD226 antigen
Gene Aliases
BC051526, DNAM-1, DNAM1, Pta1, TLiSA1
Chromosome Location
Chr. 18: 89176954 - 89272232 on Build GRCm38
UniGene ID
Mm.139293
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039149.1 NP_001034238.1 3 465
NM_178687.2 NP_848802.2 4 804
GenBank mRNA AB195429.1 - 3 577
AF416980.1 - 4 636
AF421199.1 - 3 297
AF421200.1 - 2 225
AK036455.1 - 5 899
AK140349.1 - 4 692
BC051526.1 - 3 399
BC132422.1 - 4 749 40130699
BC138769.1 - 4 749 8861791
BC145410.1 - 3 410 9053968

View Full Product Details