Gene Transcripts
- Gene Symbol
- Sh3glb2
- Entrez Gene ID
- 227700
- Gene Name
- SH3-domain GRB2-like endophilin B2
- Gene Aliases
- SPAS-1
- Chromosome Location
- Chr. 2: 30344777 - 30359316 on Build GRCm38
- UniGene ID
- Mm.33343
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001289709.1 |
NP_001276638.1
|
4, 5 |
574 |
|
NM_001289710.1 |
NP_001276639.1
|
4, 5 |
574 |
|
NM_139302.2 |
NP_647463.1
|
4, 5 |
574 |
|
GenBank mRNA |
AF426314.1 |
- |
4, 5 |
471 |
|
AK129455.1 |
- |
4, 5 |
502 |
|
AK152409.1 |
- |
4, 5 |
504 |
|
AK152502.1 |
- |
4, 5 |
525 |
|
AK155248.1 |
- |
4, 5 |
499 |
|
AK167762.1 |
- |
4, 5 |
547 |
|
BC024477.1 |
- |
4, 5 |
489 |
4977001 |
BC028859.1 |
- |
4, 5 |
563 |
4167974 |
EF676083.1 |
- |
4, 5 |
550 |
|
View Full Product Details