Gene Transcripts

Gene Symbol
PLA2G1B
Entrez Gene ID
5319
Gene Name
phospholipase A2 group IB
Gene Aliases
PLA2, PLA2A, PPLA2
Chromosome Location
Chr. 12: 120322111 - 120327789 on Build GRCh38
UniGene ID
Hs.992
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000928.2 NP_000919.1 4 533
GenBank mRNA BC005386.1 - 3 395
BC106725.1 - 4 533 40014551
BC106726.1 - 4 533 40014552
BE969737.1 - 503
BX113838.1 - 3 337
CA867923.1 - 3 376
M21054.1 - 4 534

View Full Product Details