Gene Transcripts

Gene Symbol
Tnks1bp1
Entrez Gene ID
228140
Gene Name
tankyrase 1 binding protein 1
Gene Aliases
Tab182, mKIAA1741
Chromosome Location
Chr. 2: 85048012 - 85073048 on Build GRCm38
UniGene ID
Mm.23606
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081260.2 NP_001074729.1 6 2794
GenBank mRNA AK129436.1 - 2 1487
BC025943.1 - 1 243
BC058639.1 - 2 1846
BC150947.1 - 6 2714 9087858

View Full Product Details