Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178246.3 | NP_839977.2 | 8 | 921 | |
GenBank mRNA | AK129286.1 | - | 8 | 900 | |
AK143911.1 | - | 8 | 923 | ||
AK155787.1 | - | 8 | 910 | ||
AK172217.1 | - | 7 | 801 | ||
BC024683.1 | - | 8 | 873 | 3992000 | |
BC100408.1 | - | 8 | 870 | 30734268 |