Gene Transcripts
- Gene Symbol
- Chil5
- Entrez Gene ID
- 229687
- Gene Name
- chitinase-like 5
- Gene Aliases
- Bclp, Bclp1, Bclp2, Chi3l7
- Chromosome Location
- Chr. 3: 106016919 - 106033131 on Build GRCm38
- UniGene ID
- Mm.39224
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB081756.1 |
- |
6 |
567 |
|
BC116403.1 |
- |
4 |
394 |
40102215 |
View Full Product Details