Gene Transcripts
- Gene Symbol
- Rap1gds1
- Entrez Gene ID
- 229877
- Gene Name
- RAP1, GTP-GDP dissociation stimulator 1
- Gene Aliases
- BC011279, GDS1
- Chromosome Location
- Chr. 3: 138925897 - 139075201 on Build GRCm38
- UniGene ID
- Mm.28893
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001040690.2 |
NP_001035780.1
|
6, 7 |
798 |
|
NM_001286759.1 |
NP_001273688.1
|
6, 7 |
798 |
|
NM_145544.3 |
NP_663519.2
|
5, 6 |
651 |
|
GenBank mRNA |
AK052125.1 |
- |
5, 6 |
599 |
|
AK154852.1 |
- |
6, 7 |
784 |
|
AK155793.1 |
- |
6, 7 |
784 |
|
AK160989.1 |
- |
6, 7 |
734 |
|
AK164159.1 |
- |
6, 7 |
710 |
|
AK166315.1 |
- |
6, 7 |
731 |
|
AK172052.1 |
- |
6, 7 |
800 |
|
BC011279.1 |
- |
5, 6 |
520 |
4008751 |
View Full Product Details