Gene Transcripts

Gene Symbol
Plekhn1
Entrez Gene ID
231002
Gene Name
pleckstrin homology domain containing, family N member 1
Gene Aliases
BC025458
Chromosome Location
Chr. 4: 156221456 - 156228542 on Build GRCm38
UniGene ID
Mm.423124
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008233.3 NP_001008234.2 10 1065
GenBank mRNA AK048084.1 - 8 3044
BC046544.1 - 10 1065

View Full Product Details