Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_174850.3 | NP_777275.2 | 6 | 1716 | |
GenBank mRNA | AB182579.1 | - | 6 | 1567 | |
AK044547.1 | - | 6 | 1567 | ||
AK085948.1 | - | 6 | 1687 | ||
AK151086.1 | - | 6 | 1715 | ||
AK164043.1 | - | 6 | 1687 | ||
AK165558.1 | - | 6 | 1566 | ||
AK220308.1 | - | 6 | 1493 | ||
BC043300.1 | - | 6 | 1530 | 5025620 |