Gene Transcripts

Gene Symbol
Tmf1
Entrez Gene ID
232286
Gene Name
TATA element modulatory factor 1
Gene Aliases
7030402D04Rik, Gm153, Tmf1`
Chromosome Location
Chr. 6: 97151950 - 97179124 on Build GRCm38
UniGene ID
Mm.30893
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081111.2 NP_001074580.1 16, 17 3236
GenBank mRNA BC150931.1 - 16, 17 3247

View Full Product Details