Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000303.2 | NP_000294.1 | 3 | 293 | |
GenBank mRNA | AK291537.1 | - | 3 | 264 | |
AK296123.1 | - | 3 | 286 | ||
AK300785.1 | - | 3 | 269 | ||
AK312532.1 | - | 3 | 264 | ||
BC008310.1 | - | 3 | 235 | 3611382 | |
DC366756.1 | - | 3 | 300 | ||
KU178265.1 | - | 3 | 223 | ||
U85773.1 | - | 3 | 271 |