Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_183312.3 | NP_899135.3 | 4 | 1801 | |
NM_201639.2 | NP_964001.2 | 4 | 5154 | ||
NM_207663.3 | NP_997546.2 | 5 | 4248 | ||
GenBank mRNA | AJ579700.1 | - | 4 | 5102 | |
AJ579701.1 | - | 5 | 4196 | ||
AJ579702.1 | - | 4 | 1749 | ||
AK053514.1 | - | 4 | 3266 | ||
AK143019.1 | - | 5 | 4249 | ||
AK161935.1 | - | 1 | 1731 | ||
AK172937.1 | - | 3 | 4235 | ||
BC026872.1 | - | 6 | 2316 | ||
BC040093.1 | - | 6 | 2316 | ||
BC042785.1 | - | 2 | 1231 |