Gene Transcripts

Gene Symbol
Tmem184c
Entrez Gene ID
234463
Gene Name
transmembrane protein 184C
Gene Aliases
8430433H16Rik, AI645453, AL033298, BC004056, Tmem34
Chromosome Location
Chr. 8: 77595978 - 77610759 on Build GRCm38
UniGene ID
Mm.273578
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145599.4 NP_663574.3 6 1068
GenBank mRNA AK033404.1 - 6 1037
AK164182.1 - 6 1036
BC004056.1 - 6 1026 3589819

View Full Product Details