Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177699.4 | NP_808367.2 | 19 | 3179 | |
GenBank mRNA | AB041045.1 | - | 19 | 3073 | |
AK030737.1 | - | 19 | 3141 | ||
BC017144.1 | - | 17 | 3525 | ||
BC060654.1 | - | 19 | 3162 | 30354865 |