Gene Transcripts

Gene Symbol
Aldh8a1
Entrez Gene ID
237320
Gene Name
aldehyde dehydrogenase 8 family, member A1
Gene Aliases
Raldh4
Chromosome Location
Chr. 10: 21377300 - 21396578 on Build GRCm38
UniGene ID
Mm.90181
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178713.4 NP_848828.1 5 753
GenBank mRNA AF510322.1 - 5 762
AK050298.1 - 5 754
AK143752.1 - 5 754
BC013511.1 - 3 295
BC038493.1 - 5 765 4223059

View Full Product Details