Gene Transcripts

Gene Symbol
Zdhhc22
Entrez Gene ID
238331
Gene Name
zinc finger, DHHC-type containing 22
Gene Aliases
Gm262
Chromosome Location
Chr. 12: 86980763 - 86991867 on Build GRCm38
UniGene ID
Mm.185890
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080943.2 NP_001074412.1 1 58
GenBank mRNA BC128021.1 - 1 62

View Full Product Details