Gene Transcripts

Gene Symbol
Mdc1
Entrez Gene ID
240087
Gene Name
mediator of DNA damage checkpoint 1
Gene Aliases
6820401C03, AA413496, Nfbd1, mKIAA0170
Chromosome Location
Chr. 17: 35841490 - 35859670 on Build GRCm38
UniGene ID
Mm.218511
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010833.2 NP_001010833.2 3 618
GenBank mRNA AK033009.1 - 3 455

View Full Product Details