Gene Transcripts

Gene Symbol
Mdc1
Entrez Gene ID
240087
Gene Name
mediator of DNA damage checkpoint 1
Gene Aliases
6820401C03, AA413496, Nfbd1, mKIAA0170
Chromosome Location
Chr. 17: 35841490 - 35859670 on Build GRCm38
UniGene ID
Mm.218511
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010833.2 NP_001010833.2 14 4943
GenBank mRNA AK129074.1 - 8 2679
BC085140.1 - 9 2812
BC094363.1 - 10 3646

View Full Product Details