Gene Transcripts

Gene Symbol
Exd1
Entrez Gene ID
241624
Gene Name
exonuclease 3'-5' domain containing 1
Gene Aliases
4932702D22Rik, Exdl1
Chromosome Location
Chr. 2: 119516505 - 119547627 on Build GRCm38
UniGene ID
Mm.212773
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172857.2 NP_766445.1 5, 6 1019
GenBank mRNA AK030132.1 - 5, 6 1021
BC092241.1 - 3, 4 273 30670567

View Full Product Details