Gene Transcripts

Gene Symbol
Kcng1
Entrez Gene ID
241794
Gene Name
potassium voltage-gated channel, subfamily G, member 1
Gene Aliases
AW536275, OTTMUSG00000016048
Chromosome Location
Chr. 2: 168260110 - 168283440 on Build GRCm38
UniGene ID
Mm.425102
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081134.1 NP_001074603.1 1 416
GenBank mRNA BC147251.1 - 1 348
BC147252.1 - 1 348 9056297
BC147715.1 - 1 348 9087474

View Full Product Details