Gene Transcripts
- Gene Symbol
- Pabpc4l
- Entrez Gene ID
- 241989
- Gene Name
- poly(A) binding protein, cytoplasmic 4-like
- Gene Aliases
- C330050A14Rik, EG241989
- Chromosome Location
- Chr. 3: 46442197 - 46447996 on Build GRCm38
- UniGene ID
- Mm.119234
- Species
- Mus musculus
View Full Product Details