Gene Transcripts
- Gene Symbol
- PPP2R3A
- Entrez Gene ID
- 5523
- Gene Name
- protein phosphatase 2 regulatory subunit B''alpha
- Gene Aliases
- PPP2R3, PR130, PR72
- Chromosome Location
- Chr. 3: 135965673 - 136147910 on Build GRCh38
- UniGene ID
- Hs.518155
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001190447.1 |
NP_001177376.1
|
11, 12 |
1409 |
|
NM_002718.4 |
NP_002709.2
|
12, 13 |
3844 |
|
NM_181897.2 |
NP_871626.1
|
11, 12 |
1699 |
|
XM_006713686.3 |
XP_006713749.1
|
12, 13 |
4255 |
|
XM_011512956.2 |
XP_011511258.1
|
12, 13 |
4019 |
|
XM_017006787.1 |
XP_016862276.1
|
11, 12 |
3577 |
|
GenBank mRNA |
AK293012.1 |
- |
12, 13 |
3824 |
|
AK298059.1 |
- |
11, 12 |
1338 |
|
AK299544.1 |
- |
11, 12 |
1369 |
|
AK314243.1 |
- |
12, 13 |
3772 |
|
AK316258.1 |
- |
11, 12 |
1652 |
|
AL389975.1 |
- |
9, 10 |
927 |
|
BC065531.1 |
- |
12, 13 |
3791 |
6161260 |
L07590.1 |
- |
12, 13 |
3731 |
|
L12146.1 |
- |
11, 12 |
1624 |
|
View Full Product Details