Gene Transcripts
- Gene Symbol
- Chrm2
- Entrez Gene ID
- 243764
- Gene Name
- cholinergic receptor, muscarinic 2, cardiac
- Gene Aliases
- AChR-M2, Chrm-2, M2
- Chromosome Location
- Chr. 6: 36387950 - 36528638 on Build GRCm38
- UniGene ID
- Mm.448632
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_203491.3 |
NP_987076.2
|
3 |
1564 |
|
View Full Product Details