Gene Transcripts

Gene Symbol
Nlrp4a
Entrez Gene ID
243880
Gene Name
NLR family, pyrin domain containing 4A
Gene Aliases
E330021B02, E330028A19Rik, NALP4, Nalp-eta, Nalp-ita, Nalp4a
Chromosome Location
Chr. 7: 26435113 - 26476142 on Build GRCm38
UniGene ID
Mm.322356
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172896.2 NP_766484.1 10 3043
GenBank mRNA AK054378.1 - 10 2934
AK087843.1 - 10 3044
AK135745.1 - 7 1857
AK136205.1 - 11 3019
AK162125.1 - 1 20
AY355341.1 - 10 3044
AY596200.1 - 8 2740
BC141342.1 - 10 2943 9055954
BC141343.1 - 10 2943 9055955

View Full Product Details