Gene Transcripts

Gene Symbol
Sgcz
Entrez Gene ID
244431
Gene Name
sarcoglycan zeta
Gene Aliases
C230085N17Rik
Chromosome Location
Chr. 8: 37517840 - 38661508 on Build GRCm38
UniGene ID
Mm.210388
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145841.2 NP_665840.2 2 959
GenBank mRNA AK041231.1 - 2 372
AK048960.1 - 2 923
AK136779.1 - 2 960
AY095374.1 - 1 175
AY250713.1 - 1 175
BC104048.1 - 1 175 40040143
BC104049.2 - 1 175 40040144

View Full Product Details