Gene Transcripts

Gene Symbol
Dclk3
Entrez Gene ID
245038
Gene Name
doublecortin-like kinase 3
Gene Aliases
BC056929, C730036H08, Dcamkl3
Chromosome Location
Chr. 9: 111439081 - 111489611 on Build GRCm38
UniGene ID
Mm.26361
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172928.5 NP_766516.2 2 698
GenBank mRNA AK050312.1 - 2 666
AK135010.1 - 2 687
BC056929.1 - 1 366 6816292
DQ286388.1 - 2 667

View Full Product Details