Gene Transcripts

Gene Symbol
Retnlg
Entrez Gene ID
245195
Gene Name
resistin like gamma
Gene Aliases
Fizz3, Relmg, Xcp1
Chromosome Location
Chr. 16: 48872608 - 48874498 on Build GRCm38
UniGene ID
Mm.231288
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181596.4 NP_853627.2 4 552
GenBank mRNA AB086819.1 - 3 473
AJ514933.1 - 3 476
AJ536019.1 - 4 515
AK158401.1 - 4 516

View Full Product Details