Gene Transcripts

Gene Symbol
Obox6
Entrez Gene ID
252830
Gene Name
oocyte specific homeobox 6
Gene Aliases
D17Ertd599e
Chromosome Location
Chr. 7: 15833250 - 15839679 on Build GRCm38
UniGene ID
Mm.25863
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145710.2 NP_663756.2 2 510
GenBank mRNA CN712286.1 - 2 510
CN721150.1 - 1 182

View Full Product Details