Gene Transcripts
- Gene Symbol
- Scml4
- Entrez Gene ID
- 268297
- Gene Name
- sex comb on midleg-like 4 (Drosophila)
- Gene Aliases
- 9330161D17, mFLJ00197
- Chromosome Location
- Chr. 10: 42860317 - 42960782 on Build GRCm38
- UniGene ID
- Mm.98731
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC024570.1 |
- |
4 |
615 |
|
BC043310.1 |
- |
9 |
1454 |
5067233 |
View Full Product Details