Gene Transcripts

Gene Symbol
Atp13a5
Entrez Gene ID
268878
Gene Name
ATPase type 13A5
Gene Aliases
C630015F21Rik
Chromosome Location
Chr. 16: 29231851 - 29378764 on Build GRCm38
UniGene ID
Mm.9823
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001284375.1 NP_001271304.1 10 1227
NM_175650.4 NP_783581.2 11 1362
GenBank mRNA AK083102.1 - 11 1337
AK083121.1 - 11 1363
AK158355.1 - 11 1182
BC115739.1 - 10 1160 40086065

View Full Product Details