Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023058.3 | NP_075545.2 | 3 | 1261 | |
GenBank mRNA | AF175892.1 | - | 3 | 1145 | |
AK031787.1 | - | 2 | 2128 | ||
AK158550.1 | - | 3 | 1262 | ||
AK165164.1 | - | 3 | 1128 | ||
BC025061.1 | - | 3 | 984 | 2609421 |